siRNA Details
Gene Symbol: Gm5475 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK163173.1 | - | 1 | 823 | ||
BC147350.1 | - | 1 | 503 | 9056399 | ||
BC147351.1 | - | 1 | 506 | 9056417 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
