siRNA Details
Gene Symbol: Ly6g6f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163192.1 | NP_001156664.1 | 2 | 346 | ||
GenBank mRNA | BC157908.1 | - | 2 | 346 | 9088110 | |
BC172069.1 | - | 2 | 346 | 9054355 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
