siRNA Details
Gene Symbol: Spink14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039218.2 | NP_001034307.2 | 4 | 291 | ||
GenBank mRNA | AK036889.1 | - | 1 | 2506 | ||
AV381433.2 | - | 3 | 398 | |||
BC147448.1 | - | 3 | 243 | 9056090 | ||
BC147476.1 | - | 3 | 243 | 9056118 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
