siRNA Details
Gene Symbol: Iscu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025526.4 | NP_079802.1 | 5 | 688 | ||
GenBank mRNA | AK009021.1 | - | 5 | 688 | ||
AK009421.1 | - | 5 | 692 | |||
AK039834.1 | - | 5 | 730 | |||
AK083247.1 | - | 5 | 696 | |||
BC028800.1 | - | 5 | 666 | |||
BC048409.1 | - | 5 | 685 | 6484953 | ||
BC096049.1 | - | 5 | 686 | 3466271 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
