siRNA Details
Gene Symbol: Khdc1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033904.1 | NP_001029076.1 | 1 | 388 | ||
GenBank mRNA | BC099381.1 | - | 1 | 388 | 30937206 | |
BC141037.1 | - | 82 | 9055649 |
Gene Symbol: Khdc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183322.2 | NP_899145.2 | 1 | 404 | ||
GenBank mRNA | AK136166.1 | - | 1 | 331 | ||
BY725559.1 | - | 1 | 441 |
Gene Symbol: Khdc1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113187.1 | NP_001106658.1 | 1 | 442 | ||
GenBank mRNA | AK139842.1 | - | 1 | 442 | ||
BG081161.2 | - | 1 | 168 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
