siRNA Details
Gene Symbol: Nms | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011684.2 | NP_001011684.3 | 3 | 284 | ||
GenBank mRNA | AB164466.1 | - | 3 | 284 | ||
BC138922.1 | - | 3 | 227 | 8861944 | ||
BU609935.1 | - | 3 | 230 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
