siRNA Details
Gene Symbol: Ankrd13c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013806.1 | NP_001013828.1 | 12 | 1500 | ||
GenBank mRNA | AK135906.1 | - | 13 | 1236 | ||
AK157542.1 | - | 1682 | ||||
BC065088.1 | - | 11 | 3694 | |||
BC138991.1 | - | 12 | 1508 | 8862013 | ||
BC138992.1 | - | 13 | 1487 | 8862014 | ||
CF736962.1 | - | 5 | 496 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001191570, XM_008761571, XM_006233547, XM_008763971.
Gene Ontology Categories:
