siRNA Details
Gene Symbol: Vmn2r43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198961.2 | NP_945199.1 | 1 | 191 | ||
GenBank mRNA | AJ543405.1 | - | 1 | 193 |
Gene Symbol: Vmn2r29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113468.1 | NP_001106939.1 | 1 | 151 |
Gene Symbol: Vmn2r31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105062.1 | NP_001098532.1 | 1 | 151 |
Gene Symbol: Vmn2r33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105065.2 | NP_001098535.2 | 1 | 151 |
Gene Symbol: Vmn2r35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105067.1 | NP_001098537.1 | 1 | 151 |
Gene Symbol: Vmn2r38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105070.1 | NP_001098540.1 | 1 | 151 |
Gene Symbol: Vmn2r39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105071.1 | NP_001098541.1 | 1 | 151 |
Gene Symbol: Vmn2r44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105074.1 | NP_001098544.1 | 1 | 151 |
Gene Symbol: Vmn2r46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105076.1 | NP_001098546.1 | 1 | 151 |
Gene Symbol: Vmn2r50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105178.1 | NP_001098648.1 | 1 | 151 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
