siRNA Details
Gene Symbol: Zfp141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC063263.1 | - | 7 | 1436 | ||
BC089361.1 | - | 5 | 1112 | |||
CF727770.1 | - | 1 | 563 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
