siRNA Details
Gene Symbol: Gm1966 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277179.1 | NP_001264108.1 | 3 | 1475 | ||
NM_001277180.1 | NP_001264109.1 | 3 | 1487 | |||
NM_001277181.1 | NP_001264110.1 | 4 | 1559 | |||
NM_001277182.1 | NP_001264111.1 | 4 | 1625 | |||
NM_001277183.1 | NP_001264112.1 | 6 | 1759 | |||
GenBank mRNA | AK156879.1 | - | 4 | 1541 | ||
AK171325.1 | - | 3 | 1489 | |||
AK172128.1 | - | 6 | 1737 |
