siRNA Details
Gene Symbol: Iqck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081446.1 | NP_001074915.1 | 4, 5 | 491 | ||
GenBank mRNA | AK138400.1 | - | 3, 4 | 427 | ||
AK143223.1 | - | 3, 4 | 442 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
