siRNA Details
Gene Symbol: Eif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011508.1 | NP_035638.1 | 2 | 304 | ||
GenBank mRNA | AF129888.1 | - | 2 | 289 | ||
AK148032.1 | - | 2 | 301 | |||
AK151070.1 | - | 2 | 301 | |||
AK151575.1 | - | 2 | 303 | |||
AK152194.1 | - | 2 | 303 | |||
AK169763.1 | - | 2 | 302 | |||
BC003463.1 | - | 2 | 304 | 2648657 | ||
BC010791.1 | - | 2 | 282 | 4218816 | ||
BC081429.1 | - | 2 | 291 | 5710951 | ||
BC096656.1 | - | 2 | 279 | 5143414 | ||
BC096691.1 | - | 2 | 289 | 5256396 | ||
Z50159.1 | - | 1 | 47 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001105837, FQ213689, FQ212688, FQ228394, FQ217644, and others.
Gene Ontology Categories:
