siRNA Details
Gene Symbol: Dppa5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025274.3 | NP_079550.2 | 2 | 304 | ||
GenBank mRNA | AF490349.1 | - | 2 | 260 | ||
AK010519.1 | - | 2 | 261 | |||
AK010584.1 | - | 2 | 260 | |||
AK145761.1 | - | 2 | 263 | |||
AV457404.1 | - | 2 | 306 | |||
BC092353.1 | - | 2 | 249 | 30730511 | ||
BC092354.1 | - | 2 | 257 | 30732471 | ||
BC095979.1 | - | 2 | 249 | 30732937 | ||
X57708.1 | - | 2 | 262 |
Gene Symbol: Pigp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159620.1 | NP_001153092.1 | 4 | 515 | ||
GenBank mRNA | BC061117.1 | - | 4 | 474 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
