siRNA Details
Gene Symbol: Fbxw20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008428.3 | NP_001008428.1 | 7, 8 | 1048 | ||
GenBank mRNA | AK135785.1 | - | 7, 8 | 1029 | ||
AK136200.1 | - | 7, 8 | 1026 | |||
BC085247.1 | - | 7, 8 | 1029 | 30477089 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
