siRNA Details
Gene Symbol: Ccdc13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028384.1 | NP_082660.1 | 4 | 520 | ||
GenBank mRNA | AK158659.1 | - | 2 | 300 | ||
BC147354.1 | - | 2 | 265 | 9056407 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
