siRNA Details
Gene Symbol: Gm14511 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085525.2 | NP_001078994.1 | 1 | 569 | ||
GenBank mRNA | CF106510.1 | - | 1 | 445 |
Gene Symbol: Fthl17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031261.2 | NP_112551.2 | 1 | 575 | ||
GenBank mRNA | AF285569.1 | - | 1 | 576 |
Gene Symbol: Gm14458 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099326.2 | NP_001092796.1 | 1 | 500 | ||
GenBank mRNA | BC104363.1 | - | 1 | 501 | 40049729 |
Gene Symbol: Gm5634 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085524.1 | NP_001078993.1 | 1 | 569 |
Gene Symbol: Gm5635 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038697.2 | NP_001033786.1 | 1 | 500 | ||
GenBank mRNA | BC104362.1 | - | 1 | 500 | 40049728 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
