siRNA Details
Gene Symbol: Akap14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033785.2 | NP_001028957.1 | 2 | 214 | ||
GenBank mRNA | AK133120.1 | - | 2 | 215 | ||
BC147265.1 | - | 2 | 214 | 9056322 | ||
BC147494.1 | - | 2 | 214 | 9056315 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
