siRNA Details
Gene Symbol: Rhox8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004193.2 | NP_001004193.1 | 1 | 140 | ||
GenBank mRNA | AY329370.1 | - | 1 | 140 | ||
BC139098.1 | - | 1 | 123 | 8862120 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
