siRNA Details
Gene Symbol: Arpc1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023142.2 | NP_075631.2 | 8 | 910 | ||
GenBank mRNA | AF162768.1 | - | 7 | 835 | ||
AK002274.1 | - | 8 | 911 | |||
AK005378.1 | - | 8 | 911 | |||
AK148145.1 | - | 8 | 904 | |||
AK151911.1 | - | 8 | 911 | |||
AK153510.1 | - | 8 | 911 | |||
AK157098.1 | - | 8 | 901 | |||
AK159371.1 | - | 8 | 909 | |||
AK159477.1 | - | 8 | 800 | |||
AK167978.1 | - | 8 | 840 | |||
AK168774.1 | - | 8 | 908 | |||
AK170759.1 | - | 8 | 900 | |||
AK171364.1 | - | 8 | 872 | |||
AK172124.1 | - | 8 | 878 | |||
AK215938.1 | - | 4 | 345 | |||
BC003441.1 | - | 8 | 842 | 2650641 | ||
BC010275.1 | - | 8 | 852 | 3589768 | ||
BC020404.1 | - | 5 | 438 | |||
BC092051.1 | - | 8 | 866 | 3668682 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
