siRNA Details
Gene Symbol: Ldoc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018087.1 | NP_001018097.1 | 1 | 428 | ||
GenBank mRNA | AK137057.1 | - | 1 | 538 | ||
BC052689.1 | - | 1 | 428 | 30053634 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
