siRNA Details
Gene Symbol: Gm15091 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122735.1 | NP_001116207.1 | 4, 5 | 1254 | ||
GenBank mRNA | BX513296.1 | - | 1, 2 | 33 |
Gene Symbol: Gm15127 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114400.2 | NP_001107872.2 | 4, 5 | 1248 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
