siRNA Details
Gene Symbol: RGS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102445.2 | NP_001095915.1 | 5 | 939 | ||
NM_001113380.1 | NP_001106851.1 | 4 | 514 | |||
NM_005613.5 | NP_005604.1 | 4 | 830 | |||
GenBank mRNA | AB209019.1 | - | 1 | 2208 | ||
AF493928.1 | - | 4 | 319 | |||
AK096204.1 | - | 3 | 746 | |||
AK295540.1 | - | 5 | 674 | |||
AK296240.1 | - | 4 | 358 | |||
AK300832.1 | - | 4 | 318 | |||
AK312250.1 | - | 4 | 442 | |||
BC000737.1 | - | 4 | 404 | 3510260 | ||
BC051869.1 | - | 4 | 416 | 5786695 | ||
BI547160.1 | - | 4 | 544 | |||
BT007025.1 | - | 4 | 319 | |||
DQ346662.1 | - | 4 | 443 | |||
DQ346663.1 | - | 4 | 658 | |||
DQ346664.1 | - | 5 | 770 | |||
EF054877.1 | - | 4 | 514 | |||
U27768.1 | - | 4 | 416 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
