siRNA Details
Gene Symbol: Got2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010325.2 | NP_034455.1 | 2 | 223 | ||
GenBank mRNA | AK098166.1 | - | 2 | 247 | ||
AK136556.1 | - | 2 | 260 | |||
AK147953.1 | - | 2 | 263 | |||
AK149886.1 | - | 2 | 260 | |||
AK149926.1 | - | 2 | 260 | |||
AK152921.1 | - | 2 | 260 | |||
AK155075.1 | - | 2 | 254 | |||
AK167767.1 | - | 2 | 249 | |||
AK188010.1 | - | 2 | 131 | |||
AK197746.1 | - | 2 | 131 | |||
AK204973.1 | - | 2 | 133 | |||
BC089015.1 | - | 2 | 249 | 30606379 | ||
BC089341.1 | - | 2 | 261 | 6832287 | ||
J02622.1 | - | 2 | 223 | |||
U82470.1 | - | 2 | 191 |

More Information
Other Species Targeted

Gene Ontology Categories:
