siRNA Details
Gene Symbol: Iigp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013828.2 | NP_001013850.1 | 1228 | |||
GenBank mRNA | BC020118.1 | - | 1229 | 3675329 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
