siRNA Details
Gene Symbol: Shf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013829.2 | NP_001013851.2 | 6 | 1012 | ||
GenBank mRNA | BC042839.1 | - | 6 | 900 | 4488005 | |
CB056323.1 | - | 5 | 626 |

More Information
Other Species Targeted

Gene Ontology Categories:
