siRNA Details
Gene Symbol: Gm13177 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081248.1 | NP_001074717.1 | 4 | 982 | ||
GenBank mRNA | BC150982.1 | - | 4 | 1031 | 9087895 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
