siRNA Details
Gene Symbol: Fam92b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033980.2 | NP_001029152.2 | 4 | 485 | ||
GenBank mRNA | AK131790.1 | - | 4 | 485 | ||
BC147233.1 | - | 4 | 404 | 9056283 | ||
BC147234.1 | - | 4 | 404 | 9056269 | ||
BC172014.1 | - | 3 | 169 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
