siRNA Details
Gene Symbol: Try10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038996.2 | NP_001034085.1 | 3 | 370 | ||
GenBank mRNA | AK133516.1 | - | 3 | 297 | ||
BC138698.1 | - | 3 | 370 | 8861720 | ||
BC146017.1 | - | 3 | 370 | 40131320 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
