siRNA Details
Gene Symbol: Ccin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002787.2 | NP_001002787.1 | 1 | 848 | ||
GenBank mRNA | AK030197.1 | - | 1 | 851 | ||
BC150949.1 | - | 1 | 835 | 9087860 | ||
BC150952.1 | - | 1 | 835 | 9087863 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
