siRNA Details
Gene Symbol: Znhit3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005223.2 | NP_001005223.1 | 4, 5 | 290 | ||
GenBank mRNA | AK002888.1 | - | 4, 5 | 282 | ||
AK003721.1 | - | 4, 5 | 289 | |||
AK004259.1 | - | 4, 5 | 292 | |||
AK014350.1 | - | 4, 5 | 301 | |||
BC089554.1 | - | 4, 5 | 378 | 30300471 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
