siRNA Details
Gene Symbol: Fbxl7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176959.3 | NP_795933.2 | 4 | 1725 | ||
GenBank mRNA | AK048688.1 | - | 4 | 1740 | ||
AK129227.1 | - | 4 | 1673 | |||
AK198158.1 | - | 1 | 127 | |||
BC091646.1 | - | 4 | 1725 | 30355382 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
