siRNA Details
Gene Symbol: Zfp97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011765.5 | NP_035895.3 | 4 | 1540 | ||
GenBank mRNA | L20450.1 | - | 1508 |
Gene Symbol: Zfp960 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005358.2 | NP_001005358.2 | 4 | 1405 | ||
GenBank mRNA | BC018101.1 | - | 4 | 1395 | 3494875 |
