siRNA Details
Gene Symbol: Clec4a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005860.2 | NP_001005860.1 | 3, 4 | 294 | ||
GenBank mRNA | AY397673.1 | - | 3, 4 | 294 | ||
BC104370.1 | - | 3, 4 | 294 | 40049888 | ||
BC104371.2 | - | 3, 4 | 293 |
