siRNA Details
Gene Symbol: Klri1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012520.2 | NP_001012538.2 | 5, 6 | 650 | ||
GenBank mRNA | AY324874.1 | - | 5, 6 | 650 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Klri1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012520.2 | NP_001012538.2 | 5, 6 | 650 | ||
GenBank mRNA | AY324874.1 | - | 5, 6 | 650 |