siRNA Details
Gene Symbol: Myh13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081250.1 | NP_001074719.1 | 7 | 793 | ||
GenBank mRNA | AF336977.1 | - | 5 | 334 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Myh13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081250.1 | NP_001074719.1 | 7 | 793 | ||
GenBank mRNA | AF336977.1 | - | 5 | 334 |