siRNA Details
Gene Symbol: Serpinb9g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011455.3 | NP_035585.2 | 7 | 862 | ||
GenBank mRNA | AF425083.1 | - | 7 | 857 | ||
AK005394.1 | - | 7 | 862 | |||
BC064758.1 | - | 7 | 847 | 30018322 | ||
BC064759.1 | - | 7 | 837 | 30432705 |
Gene Symbol: Serpinb9f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183197.1 | NP_899020.1 | 7 | 851 | ||
GenBank mRNA | BC052216.1 | - | 7 | 851 | 30020424 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
