siRNA Details
Gene Symbol: Gm5797 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025085.2 | NP_001020256.1 | 4 | 539 | ||
GenBank mRNA | BC090991.1 | - | 4 | 539 | 6742820 |
Gene Symbol: Gm10413 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029288.3 | NP_083564.2 | 4 | 539 | ||
GenBank mRNA | AK005583.1 | - | 4 | 529 | ||
BC100412.1 | - | 4 | 539 | 30916569 |
Gene Symbol: Gm5458 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024706.2 | NP_001019877.2 | 4 | 545 |
Gene Symbol: Gm5795 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270806.1 | NP_001257735.1 | 4 | 510 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
