siRNA Details
Gene Symbol: Cyp2d9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010006.2 | NP_034136.2 | 2 | 320 | ||
GenBank mRNA | AK143963.1 | - | 2 | 320 | ||
M23997.1 | - | 2 | 411 | |||
M23998.1 | - | 2 | 316 | |||
M27168.1 | - | 2 | 295 |
Gene Symbol: Cyp2d10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010005.3 | NP_034135.2 | 2 | 317 | ||
GenBank mRNA | BC010989.1 | - | 2 | 315 | 4193743 | |
BC051039.1 | - | 2 | 294 | |||
BC057924.1 | - | 2 | 304 | 5100459 | ||
CA457792.1 | - | 2 | 353 | |||
M27167.1 | - | 2 | 295 |
Gene Symbol: Cyp2d11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104531.1 | NP_001098001.1 | 2 | 320 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
