siRNA Details
Gene Symbol: Olfr288 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011733.2 | NP_001011733.2 | 3 | 343 | ||
GenBank mRNA | BG080619.2 | - | 3 | 343 |
Gene Symbol: Olfr287 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011780.1 | NP_001011780.1 | 3 | 404 | ||
GenBank mRNA | AK140728.1 | - | 1 | 284 |
