siRNA Details
Gene Symbol: Arsi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038499.1 | NP_001033588.1 | 2 | 738 | ||
GenBank mRNA | BC138970.1 | - | 2 | 784 | 8861992 | |
BC141169.1 | - | 2 | 784 | 9055781 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
