siRNA Details
Gene Symbol: Ms4a15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034898.2 | NP_001030070.1 | 6, 7 | 695 | ||
GenBank mRNA | AK143446.1 | - | 6, 7 | 696 | ||
BC147202.1 | - | 6, 7 | 692 | 9056250 | ||
BC172006.1 | - | 6, 7 | 756 | |||
BC172010.1 | - | 6, 7 | 770 | 9054296 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
