siRNA Details
Gene Symbol: Hmcn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024720.3 | NP_001019891.2 | 1 | 542 | ||
GenBank mRNA | AK052713.1 | - | 1 | 543 | ||
BC147549.1 | - | 1 | 410 | |||
BC147568.1 | - | 1 | 410 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
