siRNA Details
Gene Symbol: Pydc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177349.1 | NP_001170820.1 | 4 | 883 | ||
NM_001177350.1 | NP_001170821.1 | 3 | 831 | |||
GenBank mRNA | AA170499.1 | - | 2 | 365 | ||
AK143015.1 | - | 4 | 884 |
