siRNA Details
Gene Symbol: BC094916 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024721.2 | NP_001019892.2 | 4 | 849 | ||
GenBank mRNA | AK156095.1 | - | 4 | 845 | ||
AK171847.1 | - | 4 | 840 | |||
BC094916.1 | - | 4 | 743 | 30074914 | ||
JN200820.1 | - | 3 | 543 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
