siRNA Details
Gene Symbol: Gm16432 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034899.3 | NP_001030071.2 | 1, 2 | 151 | ||
GenBank mRNA | AK029617.1 | - | 151 | |||
AK131684.1 | - | 1, 2 | 76 | |||
BC147679.1 | - | 1, 2 | 151 | 9056652 | ||
BC147686.1 | - | 1, 2 | 151 | 9056659 |
