siRNA Details
Gene Symbol: Lce3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039594.1 | NP_001034683.1 | 2 | 524 | ||
GenBank mRNA | BC100538.1 | - | 2 | 524 | 30288561 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
