siRNA Details
Gene Symbol: Lce3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039594.1 | NP_001034683.1 | 2 | 457 | ||
GenBank mRNA | BC100538.1 | - | 2 | 457 | 30288561 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lce3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039594.1 | NP_001034683.1 | 2 | 457 | ||
GenBank mRNA | BC100538.1 | - | 2 | 457 | 30288561 |