siRNA Details
Gene Symbol: Pilrb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024932.2 | NP_001020103.1 | 1, 2 | 198 | ||
GenBank mRNA | AK036467.1 | - | 1, 2 | 199 | ||
BC147405.1 | - | 1, 2 | 136 | 9056472 | ||
BC147406.1 | - | 1, 2 | 136 | 9056467 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
