siRNA Details
Gene Symbol: Zar1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159693.1 | NP_001153165.1 | 3 | 706 | ||
GenBank mRNA | FJ858201.1 | - | 3 | 706 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008769045.
Gene Ontology Categories:
