siRNA Details
Gene Symbol: Tsen15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC024342.1 | - | 1, 2 | 157 | 5002659 | |
BC145701.1 | - | 1, 2 | 194 | 40131004 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
